Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine)dihydrate, Supelco®
Catalog # 85588-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
EINECS:200-754-2
CAS Number:403982-64-9
Formula:C₁₀H₂₆N₄·2H₂O