
L(+)-Glutamic acid hydrochloride for synthesis, Sigma-Aldrich®
8.14632.0250
: Merck
:
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
- Solubility:490 g/l (20 °C)
- Melting Point:191 - 193 °C decomposes
- Molar Mass:183.59 g/mol
- Bulk Density:630 kg/m3
- Density:1.525 g/cm3
:
:183.59 g/mol
:M-Clarity™ quality level = MQ200
:205-315-9
:GHS05
:Ambient
:138-15-8
:C₅H₁₀ClNO₄
:MFCD00012619