FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) 90%, pure
: 3',6'-Dihydroxy-5-isothiocyanato-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 5-FITC, Fluorescein Isothiocyanate, 5-isothiocyanatofluorescein, Fluorescein Isothiocyanate, Isomer I
:3326-32-7
:
:C₂₁H₁₁NO₅S
:MFCD00005063
:389.381 g/mol
:>360 °C
:222-042-0
:GHS08
:Refrigerator
- Environmentally Preferable:
- Appearance:Orange Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥88 %
Frequently Bought Together