Order Entry
Czech Republic
ContactUsLinkComponent
Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamide)
Toxic
Health
Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamide)
Catalog # DG024
Supplier:  G-Biosciences
CAS Number:  
Toxic
Health
Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamide)
Catalog # DG024
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :GHS06
 :GHS08
Storage Temperature:Refrigerator
CAS Number:86303-22-2
Formula:C₄₂H₇₅N₃O₁₆
MDL Number:MFCD00082541