Organic reference standard, 3,3'-Dichlorobenzidine 500 µg/ml in methanol
Katalogové číslo PROOSL20242
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:91-94-1
- Description:3,3'-Dichlorobenzidine 500 µg/ml in methanol
- Formula:C₁₂H₁₀Cl₂N₂
- Description:3,3'-Dichlorobenzidine