2,6-Diaminopurine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1904-98-9
:MFCD00071537
Formula:C₅H₆N₆
MDL Number:MFCD00213668
:150.14 g/mol
:M-Clarity™ quality level = MQ100
EINECS:217-605-2
Frequently Bought Together