Pluronic® L-61 surfactant, Sigma-Aldrich®
Catalog # SIAL435422-250ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:2000 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
CAS Number:9003-11-6
Formula:HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
MDL Number:MFCD00082049