(S)-(-)-Tetrahydro-2-furoic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:87392-07-2
:
Formula:C₅H₈O₃
MDL Number:MFCD00272639
:116.12 g/mol
ADR:8,III
:M-Clarity™ quality level = MQ100
UN:3265
Frequently Bought Together