(R)-(+)-Tetrahydro-2-furoic acid, Sigma-Aldrich®
Catalog # SIAL479292-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
ADR:8,III
:M-Clarity™ quality level = MQ100
:116.12 g/mol
UN:3265
CAS Number:87392-05-0
Formula:C₅H₈O₃
MDL Number:MFCD00211271