(Ethoxycarbonylmethyl)triphenylphosphonium bromide, Sigma-Aldrich®
Catalog # SIAL412252-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:429.30 g/mol
:M-Clarity™ quality level = MQ100
EINECS:216-230-1
CAS Number:1530-45-6
Formula:C₂₂H₂₂BrO₂P
MDL Number:MFCD00011835