(R)-(-)-N-(3,5-Dinitrobenzoyl)-α-phenylglycine, Sigma-Aldrich®
Catalog # SIAL250031-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:345.26 g/mol
CAS Number:74927-72-3
Formula:C₁₅H₁₁N₃O₇
MDL Number:MFCD00010134