Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Chymostatin
Chymostatin
Chymostatin
Catalog # ENZOALX260005M005
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Chymostatin
Catalog # ENZOALX260005M005
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
5 mg
Seq:
02
Storage conditions:
Freezer
MDL No.:
MFCD00071059
Protein/peptide name:
Chymostatin
Specifications
About this item
Product Details
Product Details