Order Entry
Czech Republic
ContactUsLinkComponent
VWR®, Aprotinin
VWR®, Aprotinin
Catalog # E429-50MG
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Aprotinin
Catalog # E429-50MG
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mg
  • CAS No.:
    9087-70-1
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00130541
  • Protein synonyms:
    Trypsin inhibitor, pancreatic basic
  • Protein/peptide name:
    Aprotinin
  • Grade:
    High purity
  • Molecular formula:
    C₂₈₄H₄₃₂N₈₄O₇₉S₇

Specifications

About this item

A competitive and reversible serine protease inhibitor of chymotrypsin, trypsin, kallikrein and plasmin. Not an inhibitor of Factor Xa and thrombin.

Activity (KIU/mg)      4500