

3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:207738-08-7
:
Formula:C₁₆H₂₀N₂·2HCl·xH₂O
MDL Number:MFCD00150104
:313.27 g/mol (anhy)
Melting Pt:>300 °C
Storage Temperature:Refrigerator
EINECS:264-769-6
UN:2811
ADR:6.1,III
:GHS07
:GHS08
Specification Test Results
- Appearance:Off-white Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥98.0 % (on dry substance)
- Water:≤12.0 % (K.F.)
Specification Test Results
Frequently Bought Together