
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71-44-3
:
Formula:C₁₀H₂₆N₄
MDL Number:MFCD00008215
:202.35 g/mol
Boiling Pt:130 °C (.1 mmHg)
Melting Pt:26…30 °C
Density:0.937 g/cm³
Flash Pt:110 °C
Storage Temperature:Refrigerator
EINECS:200-754-2
UN:3259
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to light yellow Crystalline powder or low melting mass
- Infrared spectrum:Conforms
- Melting point:26 to 30 °C
- Water:≤3 % (K.F.)
- Titration with HClO4:≥96.0 % (On dry substance)
Specification Test Results
Frequently Bought Together