Order Entry
Czech Republic
ContactUsLinkComponent
Biebrich scarlet 70% (dye content)
Biebrich scarlet 70% (dye content)
Catalog # ACRO402221000
CAS Number:  
undefined
Biebrich scarlet 70% (dye content)
Catalog # ACRO402221000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance:
    Red to brown Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤15 % (110°C)
  • Dye content:
    ≥65 %

Specification Test Results

About this item

 :
 :556.49 g/mol
Melting Pt:181…188 °C
Storage Temperature:Ambient
EINECS:224-084-5
CAS Number:4196-99-0
Formula:C₂₂H₁₄N₄Na₂O₇S₂
MDL Number:MFCD00003891