Order Entry
Czech Republic
ContactUsLinkComponent
Ammonia solution 1.7%
Irritant
Ammonia solution 1.7%
Catalog # VWRS5852.9025
Supplier:  VWR Collection
CAS Number:  
Irritant
Ammonia solution 1.7%
Catalog # VWRS5852.9025
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    25 L
  • Seq:
    5327

Specifications

About this item

 :
 :35.05 g/mol
EINECS:215-647-6
 :GHS07
Storage Temperature:Ambient
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650