Order Entry
Czech Republic
ContactUsLinkComponent
 
Hydroxypropyl methylcellulose (Hypromellose), M.W. ~86,000, 4,000 mPa.s (2% in H2O), Sigma-Aldrich®
Catalog # SIAL423203-100G
Supplier:  Merck
CAS Number:  
Hydroxypropyl methylcellulose (Hypromellose), M.W. ~86,000, 4,000 mPa.s (2% in H2O), Sigma-Aldrich®
Catalog # SIAL423203-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
CAS Number:9004-65-3
Formula:C₃H₈O₂·xCH₄O
MDL Number:MFCD00131360