Hydroxypropyl methylcellulose (Hypromellose), M.W. ~26 kDa, 80 - 120 mPa.s (2% in H2O), Millipore®
Supplier: Merck
Certificates
Hydroxypropyl methylcellulose (Hypromellose), M.W. ~26 kDa, 80 - 120 mPa.s (2% in H2O), Millipore®
Supplier: Merck
About this item
CAS Number:9004-65-3
:
Formula:C₃H₈O₂·xCH₄O
MDL Number:MFCD00131360
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach