Order Entry
Czech Republic
ContactUsLinkComponent
 
Nonidet® P 40 Substitute (NP-40) 10% in aqueous solution, Millipore®
Catalog # 492018-50
CAS Number:  
Nonidet® P 40 Substitute (NP-40) 10% in aqueous solution, Millipore®
Catalog # 492018-50
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
EINECS:500-024-6
CAS Number:9016-45-9
MDL Number:MFCD00132851