
Cetyldimethylethylammonium bromide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:124-03-8
:
Formula:C₂₀H₄₄BrN
MDL Number:MFCD00011774
:378.4741 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:204-672-8
UN:2811
:GHS07
Storage Temperature:Ambient
Frequently Bought Together