Order Entry
Czech Republic
ContactUsLinkComponent
Cetyldimethylethylammonium bromide for synthesis, Sigma-Aldrich®
Corrosive
Health
Irritant
Environment
Cetyldimethylethylammonium bromide for synthesis, Sigma-Aldrich®
Catalog # 8.18424.0100
Supplier:  Merck
CAS Number:  
Corrosive
Health
Irritant
Environment
Cetyldimethylethylammonium bromide for synthesis, Sigma-Aldrich®
Catalog # 8.18424.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    200 g/l (20 °C)
  • Melting Point:
    178 - 186 °C
  • Molar Mass:
    378.47 g/mol

Specification Test Results

About this item

 :
 :378.4741 g/mol
ADR:9,III
 :M-Clarity™ quality level = MQ200
EINECS:204-672-8
UN:3077
 :GHS05
 :GHS08
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:124-03-8
Formula:C₂₀H₄₄BrN
MDL Number:MFCD00011774