Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, Anthracene 200 µg/ml in isooctane, CIL
Organic reference standard, Anthracene 200 µg/ml in isooctane, CIL
Catalog # PROOCILULM74121.2
CAS Number:  
Organic reference standard, Anthracene 200 µg/ml in isooctane, CIL
Catalog # PROOCILULM74121.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    120-12-7
  • Description:
    Anthracene 200 µg/ml in isooctane
  • Formula:
    C₁₄H₁₀
  • Description:
    Anthracene

Specifications

About this item