Order Entry
Czech Republic
ContactUsLinkComponent
Cleanroom wipes, pre-saturated, sterile, Vertex®
Cleanroom wipes, pre-saturated, sterile, Vertex®
Supplier:  Texwipe
Certificates
undefined
Cleanroom wipes, pre-saturated, sterile, Vertex®
Supplier:  Texwipe
Certificates

Specifications

  • Catalog No:
  • 115-1890
  • 115-1410
  • 115-1412
  • Extractables - using deionised water:
  • 0,01 g/m²
  • 0,01 g/m²
  • Extractables - using isopropyl alcohol:
  • 0,02 g/m²
  • 0,02 g/m²
  • Fibres (>100 µm):
  • 200/m²
  • 200/m²
  • Particle generation (0,5 - 5 µm) - ASTM E2090 test:
  • 3,0 × 10⁶/m²
  • 3,0 × 10⁶/m²
  • Particle generation (5 - 100 µm) - ASTM E2090 Test:
  • 66000/m²
  • 66000/m²

Specifications

Frequently Bought Together