Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Earplugs
Banded Earplugs
Replacement plugs for banded earplugs, PerCap® Sperian
Replacement plugs for banded earplugs, PerCap® Sperian
Replacement plugs for banded earplugs, PerCap® Sperian
Catalog # 111-3582
Supplier:
HONEYWELL SAFETY
Expand image
Replacement plugs for banded earplugs, PerCap® Sperian
Catalog # 111-3582
Supplier:
HONEYWELL SAFETY
Specifications
Colour:
Orange
Description:
Replacement plugs for earplugs PerCap®
Environmentally Preferable:
Pk:
10 PAIR
Specifications
About this item
Product Details
Product Details