ContactUsLinkComponent
Magnetic stirring bar, set
Magnetic stirring bar, set
Catalog # HEID509-58300-00
undefined
Magnetic stirring bar, set
Catalog # HEID509-58300-00
Specifications
  • Type:
    Set of cross shape, elliptical and oval megnetic stirrer bars
  • Pk:
    10
Specifications
About this item
Set of different shaped PTFE covered magentic stirring bars.
  • Very good chemical resistance
  • Temperature resistant, suitable for high and low temperature applications
Set includes 2× each of stirrers in a plastic box, stirrers have the following shapes and dimensions, L (mm): Cross shape 10 and 16,5 mm, elliptical shape 25 mm, oval 40 and 50 mm.