ContactUsLinkComponent
Magnetic stirring bars set, cylindrical
Magnetic stirring bars set, cylindrical
Catalog # HEID509-56000-00
undefined
Magnetic stirring bars set, cylindrical
Catalog # HEID509-56000-00
Specifications
  • Type:
    Set of 3 cylindrical magnetic stirring bars
  • Pk:
    1 SET
Specifications
About this item
Set of PTFE covered magentic stirring bars.
  • Very good chemical resistance
  • Temperature resistant, suitable for high and low temperature applications
Set includes 1× each of stirrers with the following dimensions, L (mm): 25, 40 and 50 mm