
trans-Cinnamonitrile predominantly trans 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1885-38-7
:
Formula:C₉H₇N
MDL Number:MFCD00001930
:129.16 g/mol
Melting Pt:20 °C
Density:1.02 g/cm³
Flash Pt:126 °C
EINECS:224-441-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5990 to 1.6040 (20°C, 589 nm)
- GC:≥96.0 % (Sum of cis/trans)
Specification Test Results
Frequently Bought Together