Bovine alpha-Chymotrypsin type II (from Pancreas)
Dodavatel: Merck
Certifikáty
O tomto produktu
Specifikace
- CAS No.:9004-07-3
- Source:Pancreas
- Species:Bovine
- Storage conditions:Freezer
- MDL No.:MFCD00130481
- Enzyme name:alpha-Chymotrypsin type II
Specifikace
Frequently Bought Together