
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10)
Supplier: G-Biosciences
Certificates
About this item
CAS Number:15163-36-7
:
Formula:C₁₅H₃₃NO₃S
MDL Number:MFCD00036908
:307.4924 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together