4-Hydroxybenzenesulphonic acid 65% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:98-67-9
:
Formula:C₆H₆O₄S
MDL Number:MFCD00673723
:174.17 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:202-691-6
UN:1803
Frequently Bought Together