Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Lithium chloride 3 mol/l in aqueous solution
Catalog # VWRS5901.1000
Supplier:  VWR Collection
CAS Number:  
Irritant
Lithium chloride 3 mol/l in aqueous solution
Catalog # VWRS5901.1000
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :42.39 g/mol
EINECS:231-212-3
 :GHS07
Storage Temperature:Ambient
CAS Number:7447-41-8
Formula:LiCl
MDL Number:MFCD00011078