Orders LinkContactUsLinkComponent
Super Erecta® Shelving Systems, Wire
Super Erecta® Shelving Systems, Wire
Supplier:   Metro International
Super Erecta® Shelving Systems, Wire
Certificates
Super Erecta® Shelving Systems, Wire
Super Erecta® Shelving Systems, Wire
72 Options Available Below
Product Details & Documents
The open wire design of these heavy-gauge shelves minimizes dust accumulation and allows a free circulation of air, greater visibility of stored items and greater light penetration.
  • Fast, secure assembly: shelf is assembled in minutes without the use of any special tools
  • Versatile construction: by using various accessories, hundreds of shelving configurations become possible
  • Accessibility: shelves can be loaded/unloaded easily from all sides; this open construction allows use of maximum storage space of cube
  • Adjustability: shelves can be adjusted 25 mm intervals along the entire length of the post
  • Adjustable feet: bolt levelers compensate for surface irregularities
Brite: economical, chromate finish for dry, low humidity environments. Metro's Brite finish provides the look of chrome and includes a protective lacquer coat.
Chrome: the 'real' nickel-chrome finish for dry, low humidity environments. Metro's durable chrome finish includes a protective lacquer coat.
Type 304 polyshed stainless steel: addresses the most aggressive applications and environments. All-stainless solid and wire options exist for high temperature automated cart wash and autoclave applications.
Metroseal 3 with Microban: corrosion resistant finish for wet or high humidity environments with a 12 year warranty against rust and corrosion. Microban antimicrobial is built into the finish to keep the product 'cleaner between cleanings'.
Delivery information: Plastic sleeves and wedges are included with each shelf.
Product Family Options
Chrome
Product Information
  • Material
    L×W
    Weight
    Availability
    Price
  • METO1424NC
    Super Erecta® shelving system: wire shelf, 1424NC
    Super Erecta® shelving system: wire shelf, 1424NC
    Chrome plated
    610×355 mm
    2,7 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 2 500,00 Kč
    2 500,00 Kč
    1 * 1 KS
       2 500,00 Kč
  • METO1430NC
    Super Erecta® shelving system: wire shelf, 1430NC
    Super Erecta® shelving system: wire shelf, 1430NC
    Chrome plated
    760×355 mm
    3,2 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 2 620,00 Kč
    2 620,00 Kč
    1 * 1 KS
       2 620,00 Kč
  • METO1436NC
    Super Erecta® shelving system: wire shelf, 1436NC
    Super Erecta® shelving system: wire shelf, 1436NC
    Chrome plated
    914×355 mm
    3,6 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 2 620,00 Kč
    2 620,00 Kč
    1 * 1 KS
       2 620,00 Kč
  • METO1442NC
    Super Erecta® shelving system: wire shelf, 1442NC
    Super Erecta® shelving system: wire shelf, 1442NC
    Chrome plated
    1066×355 mm
    4,3 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 020,00 Kč
    3 020,00 Kč
    1 * 1 KS
       3 020,00 Kč
  • METO1448NC
    Super Erecta® shelving system: wire shelf, 1448NC
    Super Erecta® shelving system: wire shelf, 1448NC
    Chrome plated
    1219×355 mm
    4,7 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 020,00 Kč
    3 020,00 Kč
    1 * 1 KS
       3 020,00 Kč
  • METO1460NC
    Super Erecta® shelving system: wire shelf, 1460NC
    Super Erecta® shelving system: wire shelf, 1460NC
    Chrome plated
    1524×355 mm
    6,3 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 540,00 Kč
    3 540,00 Kč
    1 * 1 KS
       3 540,00 Kč
  • METO1472NC
    Super Erecta® shelving system: wire shelf, 1472NC
    Super Erecta® shelving system: wire shelf, 1472NC
    Chrome plated
    1829×355 mm
    7,7 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 980,00 Kč
    3 980,00 Kč
    1 * 1 KS
       3 980,00 Kč
  • METO1818NC
    Super Erecta® shelving system: wire shelf, 1818NC
    Super Erecta® shelving system: wire shelf, 1818NC
    Chrome plated
    457×457 mm
    2,7 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 2 730,00 Kč
    2 730,00 Kč
    1 * 1 KS
       2 730,00 Kč
  • METO1824NC
    Super Erecta® shelving system: wire shelf, 1824NC
    Super Erecta® shelving system: wire shelf, 1824NC
    Chrome plated
    610×457 mm
    3,2 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 2 730,00 Kč
    2 730,00 Kč
    1 * 1 KS
       2 730,00 Kč
  • METO1830NC
    Super Erecta® shelving system: wire shelf, 1830NC
    Super Erecta® shelving system: wire shelf, 1830NC
    Chrome plated
    760×457 mm
    3,6 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 2 770,00 Kč
    2 770,00 Kč
    1 * 1 KS
       2 770,00 Kč
  • 139-0278
    Super Erecta® shelving system: wire shelf, 1836NC
    Super Erecta® shelving system: wire shelf, 1836NC
    Chrome plated
    914×457 mm
    4,3 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 2 770,00 Kč
    2 770,00 Kč
    1 * 1 KS
       2 770,00 Kč
  • 139-0279
    Super Erecta® shelving system: wire shelf, 1842NC
    Super Erecta® shelving system: wire shelf, 1842NC
    Chrome plated
    1066×457 mm
    5,0 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 350,00 Kč
    3 350,00 Kč
    1 * 1 KS
       3 350,00 Kč
  • 139-0280
    Super Erecta® shelving system: wire shelf, 1848NC
    Super Erecta® shelving system: wire shelf, 1848NC
    Chrome plated
    1219×457 mm
    5,4
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 350,00 Kč
    3 350,00 Kč
    1 * 1 KS
       3 350,00 Kč
  • 139-0281
    Super Erecta® shelving system: wire shelf, 1854NC
    Super Erecta® shelving system: wire shelf, 1854NC
    Chrome plated
    1370×457 mm
    6,6 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 880,00 Kč
    3 880,00 Kč
    1 * 1 KS
       3 880,00 Kč
  • 139-0282
    Super Erecta® shelving system: wire shelf, 1860NC
    Super Erecta® shelving system: wire shelf, 1860NC
    Chrome plated
    1524×457 mm
    7,7 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 880,00 Kč
    3 880,00 Kč
    1 * 1 KS
       3 880,00 Kč
  • 139-0283
    Super Erecta® shelving system: wire shelf, 1872NC
    Super Erecta® shelving system: wire shelf, 1872NC
    Chrome plated
    1829×457 mm
    9,1 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 700,00 Kč
    4 700,00 Kč
    1 * 1 KS
       4 700,00 Kč
  • METO2124NC
    Super Erecta® shelving system: wire shelf, 2124NC
    Super Erecta® shelving system: wire shelf, 2124NC
    Chrome plated
    610×530 mm
    3,6 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 200,00 Kč
    3 200,00 Kč
    1 * 1 KS
       3 200,00 Kč
  • METO2130NC
    Super Erecta® shelving system: wire shelf, 2130NC
    Super Erecta® shelving system: wire shelf, 2130NC
    Chrome plated
    760×530 mm
    4,1 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 300,00 Kč
    3 300,00 Kč
    1 * 1 KS
       3 300,00 Kč
  • 139-0284
    Super Erecta® shelving system: wire shelf, 2136NC
    Super Erecta® shelving system: wire shelf, 2136NC
    Chrome plated
    914×530 mm
    5,0 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 300,00 Kč
    3 300,00 Kč
    1 * 1 KS
       3 300,00 Kč
  • 139-0285
    Super Erecta® shelving system: wire shelf, 2142NC
    Super Erecta® shelving system: wire shelf, 2142NC
    Chrome plated
    1066×530 mm
    5,4 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 840,00 Kč
    3 840,00 Kč
    1 * 1 KS
       3 840,00 Kč
  • 139-0286
    Super Erecta® shelving system: wire shelf, 2148NC
    Super Erecta® shelving system: wire shelf, 2148NC
    Chrome plated
    1219×530 mm
    6,4
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 840,00 Kč
    3 840,00 Kč
    1 * 1 KS
       3 840,00 Kč
  • 139-0287
    Super Erecta® shelving system: wire shelf, 2154NC
    Super Erecta® shelving system: wire shelf, 2154NC
    Chrome plated
    1370×530 mm
    7,3 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 580,00 Kč
    4 580,00 Kč
    1 * 1 KS
       4 580,00 Kč
  • 139-0288
    Super Erecta® shelving system: wire shelf, 2160NC
    Super Erecta® shelving system: wire shelf, 2160NC
    Chrome plated
    1524×530 mm
    8,2 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 580,00 Kč
    4 580,00 Kč
    1 * 1 KS
       4 580,00 Kč
  • 139-0289
    Super Erecta® shelving system: wire shelf, 2172NC
    Super Erecta® shelving system: wire shelf, 2172NC
    Chrome plated
    1829×530 mm
    10,9 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 5 430,00 Kč
    5 430,00 Kč
    1 * 1 KS
       5 430,00 Kč
  • METO2424NC
    Super Erecta® shelving system: wire shelf, 2424NC
    Super Erecta® shelving system: wire shelf, 2424NC
    Chrome plated
    610×610 mm
    4,1 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 390,00 Kč
    3 390,00 Kč
    1 * 1 KS
       3 390,00 Kč
  • 139-0290
    Super Erecta® shelving system: wire shelf, 2436NC
    Super Erecta® shelving system: wire shelf, 2436NC
    Chrome plated
    914×610 mm
    5,9 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 550,00 Kč
    3 550,00 Kč
    1 * 1 KS
       3 550,00 Kč
  • 139-0291
    Super Erecta® shelving system: wire shelf, 2442NC
    Super Erecta® shelving system: wire shelf, 2442NC
    Chrome plated
    1066×610 mm
    6,8 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 120,00 Kč
    4 120,00 Kč
    1 * 1 KS
       4 120,00 Kč
  • 139-0292
    Super Erecta® shelving system: wire shelf, 2448NC
    Super Erecta® shelving system: wire shelf, 2448NC
    Chrome plated
    1219×610 mm
    7,3 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 120,00 Kč
    4 120,00 Kč
    1 * 1 KS
       4 120,00 Kč
  • 139-0293
    Super Erecta® shelving system: wire shelf, 2454NC
    Super Erecta® shelving system: wire shelf, 2454NC
    Chrome plated
    1370×610 mm
    8,6 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 890,00 Kč
    4 890,00 Kč
    1 * 1 KS
       4 890,00 Kč
  • 139-0294
    Super Erecta® shelving system: wire shelf, 2460NC
    Super Erecta® shelving system: wire shelf, 2460NC
    Chrome plated
    1524×610 mm
    9,5 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 890,00 Kč
    4 890,00 Kč
    1 * 1 KS
       4 890,00 Kč
  • 139-0295
    Super Erecta® shelving system: wire shelf, 2472NC
    Super Erecta® shelving system: wire shelf, 2472NC
    Chrome plated
    1829×610 mm
    11,8 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 5 870,00 Kč
    5 870,00 Kč
    1 * 1 KS
       5 870,00 Kč
  • METO3060NC
    Super Erecta® shelving system: wire shelf, 3060NC
    Super Erecta® shelving system: wire shelf, 3060NC
    Chrome plated
    1524×760 mm
    11,8 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 6 620,00 Kč
    6 620,00 Kč
    1 * 1 KS
       6 620,00 Kč
  • METO3072NC
    Super Erecta® shelving system: wire shelf, 3072NC
    Super Erecta® shelving system: wire shelf, 3072NC
    Chrome plated
    1829×760 mm
    14,0 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 7 900,00 Kč
    7 900,00 Kč
    1 * 1 KS
       7 900,00 Kč
  • METO3648NC
    Super Erecta® shelving system: wire shelf, 3648NC
    Super Erecta® shelving system: wire shelf, 3648NC
    Chrome plated
    1219×910 mm
    10,4 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 6 940,00 Kč
    6 940,00 Kč
    1 * 1 KS
       6 940,00 Kč
Stainless steel
Product Information
  • Material
    L×W
    Weight
    Availability
    Price
  • METO1424NS
    Super Erecta® shelving system: wire shelf, 1424NS
    Super Erecta® shelving system: wire shelf, 1424NS
    Stainless steel
    610×355 mm
    2,7 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 7 770,00 Kč
    7 770,00 Kč
    1 * 1 KS
       7 770,00 Kč
  • METO1430NS
    Super Erecta® shelving system: wire shelf, 1430NS
    Super Erecta® shelving system: wire shelf, 1430NS
    Stainless steel
    760×355 mm
    3,2 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 8 090,00 Kč
    8 090,00 Kč
    1 * 1 KS
       8 090,00 Kč
  • METO1436NS
    Super Erecta® shelving system: wire shelf, 1436NS
    Super Erecta® shelving system: wire shelf, 1436NS
    Stainless steel
    914×355 mm
    3,6 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 7 530,00 Kč
    7 530,00 Kč
    1 * 1 KS
       7 530,00 Kč
  • METO1460NS
    Super Erecta® shelving system: wire shelf, 1460NS
    Super Erecta® shelving system: wire shelf, 1460NS
    Stainless steel
    1524×355 mm
    6,3 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 12 480,00 Kč
    12 480,00 Kč
    1 * 1 KS
       12 480,00 Kč
  • METO1472NS
    Super Erecta® shelving system: wire shelf, 1472NS
    Super Erecta® shelving system: wire shelf, 1472NS
    Stainless steel
    1829×355 mm
    7,7 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 14 030,00 Kč
    14 030,00 Kč
    1 * 1 KS
       14 030,00 Kč
  • METO1824NS
    Super Erecta® shelving system: wire shelf, 1824NS
    Super Erecta® shelving system: wire shelf, 1824NS
    Stainless steel
    610×457 mm
    3,2 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 9 790,00 Kč
    9 790,00 Kč
    1 * 1 KS
       9 790,00 Kč
  • 139-0296
    Super Erecta® shelving system: wire shelf, 1836NS
    Super Erecta® shelving system: wire shelf, 1836NS
    Stainless steel
    914×457 mm
    4,3 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 9 890,00 Kč
    9 890,00 Kč
    1 * 1 KS
       9 890,00 Kč
  • 139-0297
    Super Erecta® shelving system: wire shelf, 1842NS
    Super Erecta® shelving system: wire shelf, 1842NS
    Stainless steel
    1066×457 mm
    5,0 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 11 720,00 Kč
    11 720,00 Kč
    1 * 1 KS
       11 720,00 Kč
  • 139-0298
    Super Erecta® shelving system: wire shelf, 1848NS
    Super Erecta® shelving system: wire shelf, 1848NS
    Stainless steel
    1219×457 mm
    5,4 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 11 720,00 Kč
    11 720,00 Kč
    1 * 1 KS
       11 720,00 Kč
  • 139-0308
    Super Erecta® shelving system: wire shelf, 2148NS
    Super Erecta® shelving system: wire shelf, 2148NS
    Stainless steel
    1219×457 mm
    6,4 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 13 180,00 Kč
    13 180,00 Kč
    1 * 1 KS
       13 180,00 Kč
  • 139-0299
    Super Erecta® shelving system: wire shelf, 1854NS
    Super Erecta® shelving system: wire shelf, 1854NS
    Stainless steel
    1370×457 mm
    6,6 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 14 870,00 Kč
    14 870,00 Kč
    1 * 1 KS
       14 870,00 Kč
  • 139-0304
    Super Erecta® shelving system: wire shelf, 1860NS
    Super Erecta® shelving system: wire shelf, 1860NS
    Stainless steel
    1524×457 mm
    7,7 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 14 870,00 Kč
    14 870,00 Kč
    1 * 1 KS
       14 870,00 Kč
  • 139-0305
    Super Erecta® shelving system: wire shelf, 1872NS
    Super Erecta® shelving system: wire shelf, 1872NS
    Stainless steel
    1829×457 mm
    9,1 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 17 670,00 Kč
    17 670,00 Kč
    1 * 1 KS
       17 670,00 Kč
  • METO2124NS
    Super Erecta® shelving system: wire shelf, 2124NS
    Super Erecta® shelving system: wire shelf, 2124NS
    Stainless steel
    610×530 mm
    3,6 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 11 370,00 Kč
    11 370,00 Kč
    1 * 1 KS
       11 370,00 Kč
  • METO2130NS
    Super Erecta® shelving system: wire shelf, 2130NS
    Super Erecta® shelving system: wire shelf, 2130NS
    Stainless steel
    760×530 mm
    4,1 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 10 680,00 Kč
    10 680,00 Kč
    1 * 1 KS
       10 680,00 Kč
  • 139-0306
    Super Erecta® shelving system: wire shelf, 2136NS
    Super Erecta® shelving system: wire shelf, 2136NS
    Stainless steel
    914×530 mm
    5,0 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 11 480,00 Kč
    11 480,00 Kč
    1 * 1 KS
       11 480,00 Kč
  • 139-0307
    Super Erecta® shelving system: wire shelf, 2142NS
    Super Erecta® shelving system: wire shelf, 2142NS
    Stainless steel
    1066×530 mm
    5,4 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 13 180,00 Kč
    13 180,00 Kč
    1 * 1 KS
       13 180,00 Kč
  • 139-0309
    Super Erecta® shelving system: wire shelf, 2154NS
    Super Erecta® shelving system: wire shelf, 2154NS
    Stainless steel
    1370×530 mm
    7,3 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 17 480,00 Kč
    17 480,00 Kč
    1 * 1 KS
       17 480,00 Kč
  • 139-0310
    Super Erecta® shelving system: wire shelf, 2160NS
    Super Erecta® shelving system: wire shelf, 2160NS
    Stainless steel
    1524×530 mm
    8,2 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 17 480,00 Kč
    17 480,00 Kč
    1 * 1 KS
       17 480,00 Kč
  • 139-0311
    Super Erecta® shelving system: wire shelf, 2172NS
    Super Erecta® shelving system: wire shelf, 2172NS
    Stainless steel
    1829×530 mm
    10,9 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 20 670,00 Kč
    20 670,00 Kč
    1 * 1 KS
       20 670,00 Kč
  • 139-0312
    Super Erecta® shelving system: wire shelf, 2436NS
    Super Erecta® shelving system: wire shelf, 2436NS
    Stainless steel
    914×610 mm
    5,9 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 12 200,00 Kč
    12 200,00 Kč
    1 * 1 KS
       12 200,00 Kč
  • 139-0313
    Super Erecta® shelving system: wire shelf, 2442NS
    Super Erecta® shelving system: wire shelf, 2442NS
    Stainless steel
    1066×610 mm
    6,8 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 14 730,00 Kč
    14 730,00 Kč
    1 * 1 KS
       14 730,00 Kč
  • 139-0314
    Super Erecta® shelving system: wire shelf, 2448NS
    Super Erecta® shelving system: wire shelf, 2448NS
    Stainless steel
    1219×610 mm
    7,3 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 14 730,00 Kč
    14 730,00 Kč
    1 * 1 KS
       14 730,00 Kč
  • 139-0315
    Super Erecta® shelving system: wire shelf, 2454NS
    Super Erecta® shelving system: wire shelf, 2454NS
    Stainless steel
    1370×610 mm
    8,6 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 17 950,00 Kč
    17 950,00 Kč
    1 * 1 KS
       17 950,00 Kč
  • 139-0316
    Super Erecta® shelving system: wire shelf, 2460NS
    Super Erecta® shelving system: wire shelf, 2460NS
    Stainless steel
    1524×610 mm
    9,5 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 17 950,00 Kč
    17 950,00 Kč
    1 * 1 KS
       17 950,00 Kč
  • 139-0317
    Super Erecta® shelving system: wire shelf, 2472NS
    Super Erecta® shelving system: wire shelf, 2472NS
    Stainless steel
    1829×610 mm
    11,8 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 22 320,00 Kč
    22 320,00 Kč
    1 * 1 KS
       22 320,00 Kč
  • METO3048NS
    Super Erecta® shelving system: wire shelf, 3048NS
    Super Erecta® shelving system: wire shelf, 3048NS
    Stainless steel
    1219×760 mm
    9,5 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 19 470,00 Kč
    19 470,00 Kč
    1 * 1 KS
       19 470,00 Kč
Metroseal 3 with Microban
Product Information
  • Material
    L×W
    Weight
    Availability
    Price
  • METO1460NK3
    Super Erecta® shelving system: wire shelf, 1460NK3
    Super Erecta® shelving system: wire shelf, 1460NK3
    Metroseal 3
    1524×355 mm
    6,3 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 890,00 Kč
    3 890,00 Kč
    1 * 1 KS
       3 890,00 Kč
  • METO1824NK3
    Super Erecta® shelving system: wire shelf, 1824NK3
    Super Erecta® shelving system: wire shelf, 1824NK3
    Metroseal 3
    610×457 mm
    3,2 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 190,00 Kč
    3 190,00 Kč
    1 * 1 KS
       3 190,00 Kč
  • METO1830NK3
    Super Erecta® shelving system: wire shelf, 1830NK
    Super Erecta® shelving system: wire shelf, 1830NK
    Metroseal 3
    760×457 mm
    3,6 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 370,00 Kč
    3 370,00 Kč
    1 * 1 KS
       3 370,00 Kč
  • METO1836NK3
    Super Erecta® shelving system: wire shelf, 1836NK3
    Super Erecta® shelving system: wire shelf, 1836NK3
    Metroseal 3
    914×457 mm
    4,3 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 370,00 Kč
    3 370,00 Kč
    1 * 1 KS
       3 370,00 Kč
  • METO1842NK3
    Super Erecta® shelving system: wire shelf, 1842NK3
    Super Erecta® shelving system: wire shelf, 1842NK3
    Metroseal 3
    1066×457 mm
    5,0 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 020,00 Kč
    4 020,00 Kč
    1 * 1 KS
       4 020,00 Kč
  • METO1848NK3
    Super Erecta® shelving system: wire shelf, 1848NK3
    Super Erecta® shelving system: wire shelf, 1848NK3
    Metroseal 3
    1219×457 mm
    5,4 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 020,00 Kč
    4 020,00 Kč
    1 * 1 KS
       4 020,00 Kč
  • METO1854NK3
    Super Erecta® shelving system: wire shelf, 1854NK3
    Super Erecta® shelving system: wire shelf, 1854NK3
    Metroseal 3
    1370×457 mm
    6,6 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 740,00 Kč
    4 740,00 Kč
    1 * 1 KS
       4 740,00 Kč
  • METO1860NK3
    Super Erecta® shelving system: wire shelf, 1860NK3
    Super Erecta® shelving system: wire shelf, 1860NK3
    Metroseal 3
    1524×457 mm
    7,7 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 740,00 Kč
    4 740,00 Kč
    1 * 1 KS
       4 740,00 Kč
  • METO2442NK3
    Super Erecta® shelving system: wire shelf, 2442NK3
    Super Erecta® shelving system: wire shelf, 2442NK3
    Metroseal 3
    1066×610 mm
    6,8 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 950,00 Kč
    4 950,00 Kč
    1 * 1 KS
       4 950,00 Kč
  • METO2460NK3
    Super Erecta® shelving system: wire shelf, 2460NK3
    Super Erecta® shelving system: wire shelf, 2460NK3
    Metroseal 3
    1524×610 mm
    9,5 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 5 870,00 Kč
    5 870,00 Kč
    1 * 1 KS
       5 870,00 Kč
  • METO2472NK3
    Super Erecta® shelving system: wire shelf, 2472NK3
    Super Erecta® shelving system: wire shelf, 2472NK3
    Metroseal 3
    1829×610 mm
    11,8 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 6 920,00 Kč
    6 920,00 Kč
    1 * 1 KS
       6 920,00 Kč