Ensure optimal security for critical samples by using Thermo Scientific™ CO₂ and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
- LN₂ backup system maintains temperature down to –80 °C with liquid nitrogen, CO₂ backup system maintains temperature down to –67 °C with CO₂
LN₂ backup system: With LN₂ emergency cooling, a mobile tank is sufficient if it is ensured that the fill level is regularly replenished. The supply pressure must be less than ~2,5 bar, this is usually given directly in the case of mobile tanks, and an upstream LN₂ pressure reducer is required for central ring lines which are operated in the order of 4 bar.
CO₂ backup system: Bottles/cylinder batteries or large-capacity tanks are possible for the CO₂ supply, but it must be ensured that the CO₂ is removed from the liquid phase (riser tube bottles).
- Catalog No:
- HERAFCO300A
- 471-1232
- HERALNTSXU
- 471-1234
- HERAFLN300W
- 471-1233
- HERAFLN4567
- HERAFLN300D
- HERAFCO300D
- HERAFLN300A
- 471-1235
- HERALN4567
- HERAFCO4567
- HERAFCO300W
- Cat. No.:
- HERAFCO300A
- 471-1232
- 471-1234
- HERAFLN300W
- 471-1233
- HERAFLN4567
- HERAFLN300D
- HERAFCO300D
- HERAFLN300A
- 471-1235
- HERALN4567
- HERAFCO4567
- HERAFCO300W
- Electrical:
- 115 V/60 Hz
- —
- 208-230 V/60 Hz
- 230 V/50 Hz
- —
- 230 V/50 Hz