Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
Tetracycline hydrochloride can be used as secondary standard
 
undefined
 
 
Tetracycline hydrochloride can be used as secondary standard

 

  • Appearance:
    Yellow Powder
  • Infrared spectrum:
    Conforms
  • Purity:
    ≥80 % (as is, calculated)

 

 

Frequently Bought Together