Order Entry
Czech Republic
ContactUsLinkComponent
Bradford's reagent for quantitative protein determination or 0,1 - 1,4 mg/ml protein, Supelco®
Corrosive
Health
Bradford's reagent for quantitative protein determination or 0,1 - 1,4 mg/ml protein, Supelco®
Catalog # SIALB6916-500ML
Supplier:  Merck
CAS Number:  
Corrosive
Health
Bradford's reagent for quantitative protein determination or 0,1 - 1,4 mg/ml protein, Supelco®
Catalog # SIALB6916-500ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance (Color):
    Faint Yellow-Brown to Light Brown
  • Appearance (Form):
    Solution
  • Suitability, Suitable for protein determination:
    Suitable

Specification Test Results

About this item

 :
ADR:8,III
 :M-Clarity™ quality level = MQ200
UN:1805
 :GHS05
 :GHS08