Order Entry
Czech Republic
ContactUsLinkComponent
 
Health
Bradford's reagent for quantitative protein determination
Catalog # A6932.0500
Supplier:  PanReac AppliChem
CAS Number:  
Health
Bradford's reagent for quantitative protein determination
Catalog # A6932.0500
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :GHS08
Storage Temperature:Refrigerator