Pierce™ Coomassie Plus (Bradford) Protein Assay Kit
PIER23236
:
- Pk:1 KIT
- Description:Coomassie Plus™ (Bradford) Protein Assay Kit
The Pierce™ Coomassie Plus Protein assay is a quick and ready-to-use, reducing agent-compatible, improved Bradford assay to quickly measure (A595 nm) total protein concentration compared to a protein standard.
- Colorimetric measure with a standard spectrophotometer or plate reader (595 nm)
- Easy to use single reagent; no working reagent preparation required
- Fast almost immediate colour development; add, mix and read results
- Broad range detects protein concentration in the range 1 to 1500 µg/ml
- Better improved linearity and response-uniformity compared to traditional Bradford formulations
- Flexible microplate and cuvette protocols provided with the instructions and adaptable to several target working ranges
This modification of the well-known Bradford method greatly reduces the tendency of Coomassie reagents to give non linear response curves by a formulation that substantially improves linearity for a defined range of protein concentration. In addition, the Coomassie Plus Reagent results in significantly less protein-to-protein variation than is observed with other Bradford-type Coomassie formulations.
: Kit contains sufficient reagents for 630 tube assays or 3160 microplate assays. Pierce™ Coomassie Plus™ Protein Assay Reagent, 950 ml; Albumin Standard Ampoules, 2 mg/ml, 10×1 ml.