Order Entry
Czech Republic
ContactUsLinkComponent
Buffer, TRIS-glycine solution 10X concentrate (TG buffer), Sigma-Aldrich®
Buffer, TRIS-glycine solution 10X concentrate (TG buffer), Sigma-Aldrich®
Catalog # SIAL93015-10L-F
Supplier:  Merck
CAS Number:  
Buffer, TRIS-glycine solution 10X concentrate (TG buffer), Sigma-Aldrich®
Catalog # SIAL93015-10L-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 L
  • Description:
    TRIS-glycine buffer solution 10X concentrate (TG buffer)

Specifications

About this item