Order Entry
Czech Republic
ContactUsLinkComponent
Buffer, SSC solution 20X concentrate (saline-sodium citrate), Sigma-Aldrich®
Buffer, SSC solution 20X concentrate (saline-sodium citrate), Sigma-Aldrich®
Catalog # SIALS6639-1L
Supplier:  Merck
CAS Number:  
Buffer, SSC solution 20X concentrate (saline-sodium citrate), Sigma-Aldrich®
Catalog # SIALS6639-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    SSC buffer solution 20X concentrate (saline-sodium citrate)
  • M-Clarity quality level:
    MQ200

Specifications

About this item