Buffer, SSC solution 20X concentrate (saline-sodium citrate), Sigma-Aldrich®
Katalogové číslo SIAL93017-10L-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 L
- Description:SSC buffer solution 20X concentrate (saline-sodium citrate)