ContactUsLinkComponent
VWR®, Nucleic acid loading dyes
VWR®, Nucleic acid loading dyes
Catalog # E190-5ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Nucleic acid loading dyes
Catalog # E190-5ML
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    5 mL
  • Description:
    Agarose gel loading dye with 3 tracking dyes and 15% Ficoll (6X)
Specifications
About this item
Designed to optimise loading nucleic acids in agarose gels, the loading dye imparts colour to the sample to facilitate the loading process and increase the density of the sample to ensure efficient sample distribution into each well. The dye migrates independently from the sample, allowing estimation of the migration of nucleic acids.
  • Eliminates exposure to irritating and harmful powdered dyes
  • Free from DNase, RNase and protease
  • Functionally and analytically tested