
Pyrrole-3-carboxylic acid hydrate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:336100-46-0
:
Formula:C₅H₅NO₂·H₂O
MDL Number:MFCD06201862
Melting Pt:144…147 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Beige to brown Crystalline powder
- Infrared spectrum:Conforms
- Water:≤14 %
- HPLC:≥94 %
- Titration with NaOH:≥94 %
Specification Test Results
Frequently Bought Together