Herbicide reference standard, Hexazinone 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA14200000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:51235-04-2
- Description:Hexazinone 100 µg/ml in acetonitrile
- Formula:C₁₂H₂₀N₄O₂
- Description:Hexazinone