Phosphamidon (mixture of cis and trans isomers)
Katalogové číslo EHERC16140000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
EINECS:236-116-5
UN:2810
ADR:6.1,III
CAS Number:13171-21-6
Formula:C₁₀H₁₉ClNO₅P