(2-Hydroxylpropyl)-β-cyclodextrine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:128446-35-5
:MFCD00069372
Formula:C₆₃H₁₁₂O₄₂
MDL Number:MFCD16621721
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
Frequently Bought Together