

Malathion (1,2-bis(ethoxycarbonyl)ethyl-O,O-dimethyl phosphorodithioate)
Catalog # PROOLGC1205
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
EINECS:204-497-7
UN:3082
ADR:9,III
:GHS07
:GHS09
CAS Number:121-75-5
Formula:C₁₀H₁₉O₆PS₂