Malathion (1,2-Bis(ethoxycarbonyl)ethyl O,O-dimethyl phosphorodithioate)
Catalog # EHERC14710000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
EINECS:204-497-7
CAS Number:121-75-5
Formula:C₁₀H₁₉O₆PS₂