
4-(Benzyloxy)-3-methoxybenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2426-87-1
:
Formula:C₁₅H₁₄O₃
MDL Number:MFCD00003365
:242.27 g/mol
Boiling Pt:213…215 °C (3.5 mmHg)
Melting Pt:61…65 °C
Storage Temperature:Ambient
EINECS:219-379-0
:GHS07
Related Information
Frequently Bought Together