
3-(Methoxycarbonyl)phenylboronic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99769-19-4
:
Formula:C₈H₉BO₄
MDL Number:MFCD02093046
:179.97 g/mol
Melting Pt:136 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together